Large Magnetic Demonstration Prism Set
Catalog # 470002-942
Supplier: United Scientific Supplies
Specifications
- Description:Large Magnetic Demonstration Prism Set
- Cat. No.:470002-942
- Supplier no.:PST620
Specifications
About this item
Teach a wide variety of optical principles with this set of seven lenses and prisms
- Durable storage box
- Magnetic backing on optical components
An oversized set of 6 lenses and prisms designed for mounting on a magnetic chalk or marker board. Lenses are about 8" long, so your optics experiments would be clearly visible from every part of the classroom. Each piece in the set has a pre-attached magnetic backing. An activity guide is also included. Set comes in a sturdy wooden storage case. Please note: Laser Ray Box not included.
Ordering information: Contents include Semi-circular lens; Equilateral prism, 75mm sides; Double convex lens; Double concave lens; Single convex lens; and Single concave lens.