Center of Mass Apparatus
470110-826
- Description:Center of Mass Apparatus
- Cat. No.:470110-826
- Supplier no.:18650
Illustrates How Two Bodies Rotate Freely About their Common Center of Mass
- Includes rod with mounted spheres, wooden handle with pin and instructions
A 54cm rod supports two spheres of different mass at its ends. The rod has two poles, one at its geometric center and one at the model’s center of mass. Mounted at one hole the system spins freely; at the other it does not.