Ward's® Cat or Rabbit Blood Slide
470177-122
: Avantor
- Description:Blood slide
- Environmentally Preferable:
- Cat. No.:470177-122
- Stain:Giemsa
- Supplier no.:WARD470177-122
- Orientation:Smear
Illustrates characteristics of blood cell types.
- Identifying features clearly distinguishable
- Rigorous quality control standards
The red blood cells are distinctly smaller than human red blood cells.