Bullseye® Compass by SAFE-T®
Catalog # 470002-826
Supplier: Learning Resources
Specifications
- Description:Bullseye® compass, 6"
- Cat. No.:470002-826
- Supplier no.:45700
Specifications
About this item
SAFE-T® Plastic for a safer and more durable instrument.
- Built-in protractor both measures and constructs angles
The opening in the Direct View™ compass point provides for the easiest, most accurate location for all circle constructions. Quick-Set™ radius indicator allows immediate comparison of inches and centimeters and setting adjustments without removing the compass from the drawing surface.