Boyle’s Law Apparatus
470110-858
: Avantor
- Description:Boyle’s Law Apparatus
- Cat. No.:470110-858
- Supplier no.:1080-90
This low-cost apparatus can demonstrate the concept of Boyle’s Law easily and safely.
- Simple for Students to Use
- Low-Cost
- No Mercury Needed
- Instructions Provided
- Plot of Depth vs. Air Verifies Boyle's Law
No mercury is needed. Just fill the outer cylinder 2/3 full of water and invert the inner cylinder into the outer. The plot of depth versus air that develops by reading the depth of the inner cylinder in the water and the air level in the inner cylinder verifies Boyle’s Law.
This apparatus includes instructions.