ContactUsLinkComponent
Blunt-Point Surgical Scissors
Blunt-Point Surgical Scissors
Catalog # 470018-946
undefined
Blunt-Point Surgical Scissors
Catalog # 470018-946
Supplier Number:  140985
Specifications
  • Description:
    Blunt-Point Surgical Scissors
  • Length:
    140 mm (5.5")
  • Material:
    Stainless steel
  • Cat. No.:
    470018-946
  • Supplier no.:
    140985
  • Blade type:
    Blunt/Blunt
Specifications
About this item
When working with particularly delicate specimens, the blunt tips of these straight scissors reduce the chance of tearing, damaging, or puncturing tissue adjacent to the cut. Stainless steel.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.