ContactUsLinkComponent
CENCO® AP Physics Lab 16: Mapping Electric Fields
CENCO® AP Physics Lab 16: Mapping Electric Fields
Catalog # 470018-140
Supplier:  Avantor
undefined
CENCO® AP Physics Lab 16: Mapping Electric Fields
Catalog # 470018-140
Supplier:  Avantor
Specifications
  • Description:
    CENCO AP Physics Lab 16: Mapping Electric Fields
  • Environmentally Preferable:
  • Cat. No.:
    470018-140
  • Supplier no.:
    WARD470018-140
Specifications
About this item
In this lab, students create maps of the electric field that exists between two charged surfaces: circular “points” or plates.
  • College Board AP Curriculum-Aligned Labs
  • Students Create Maps of Electric Field between Two Charged Surfaces
  • Material and Instructions for Six Students per Lab Group
  • Kit Includes Cork Board Surface, Electrical Conductive Paper, and More
The kit contains instructions and materials included a cork board surface, electrical conductive paper, and more, so teachers can conduct the lab without searching for materials.
The lab is aligned with the College Board AP Curriculum.
Accessories information: AP Physics instructors authored the teacher's guides and student manuals, which are available separately.
Ordering information: The kit includes a cork board surface, electrical conductive paper, conductive ink pen, circular template, and a multimeter.