Orders LinkContactUsLinkComponent
Aorta Slide
Aorta Slide
Aorta Slide, Section, Verhoeff and Eosin
Catalog # 470183-006
Supplier:   Avantor
Aorta Slide
Catalog # 470183-006
Supplier:  Avantor
Aorta Slide
Aorta Slide
Aorta Slide, Section, Verhoeff and Eosin
Price
$10.00
Log in for availability
Each
Product Details & Documents
Observe elastic tissues within the aorta.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Human. Elastic tissue stained.
Specifications
  • Description:
    Aorta Slide
  • Stain:
    Verhoeff and Eosin
  • Orientation:
    Section