Orders LinkContactUsLinkComponent
Amphioxus, Composite Slide
Amphioxus, Composite Slide
Amphioxus, Composite, Cross section, Mallory Triple
Catalog # 470181-690
Supplier:   Avantor
Amphioxus, Composite Slide
Catalog # 470181-690
Supplier:  Avantor
Amphioxus, Composite Slide
Amphioxus, Composite Slide
Amphioxus, Composite, Cross section, Mallory Triple
Price
$26.95
Log in for availability
Each
Product Details & Documents
Lancelet Study
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Cross section through the buccal funnel, anterior pharynx, gonads, intestine, and tail.
Specifications
  • Description:
    Amphioxus, Composite
  • Stain:
    Mallory Triple
  • Orientation:
    Cross section