Ammonium Molybdate
Product Details & Documents
CAS Number: 12054-85-2
Formula Weight: 1235.86
Formula: (NH4)6Mo7O24·4H2O
Hazard Info: Irritant
Density (g/mL): 2.276
Boiling Point (°C): Decomposes
Freezing Point (°C): Decomposes
Synonyms: Ammonium Heptamolybdate Tetrahydrate
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Documents
Specifications
Specifications
- Description:Ammonium Molybdate 4-Hydrate
- Size:100 g
- Chemical/physical specifications:Crystal
- Grade:Reagent
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



