Orders LinkContactUsLinkComponent
Adjustable Right Angle Clamp Holder
Adjustable Right Angle Clamp Holder
Adjustable Right Angle Clamp Holder
Catalog # 89427-302
Supplier #:  H-8342
Adjustable Right Angle Clamp Holder
Catalog # 89427-302
Adjustable Right Angle Clamp Holder
Adjustable Right Angle Clamp Holder
Adjustable Right Angle Clamp Holder
Price
$34.53
Log in for availability
Each
Product Details & Documents
Sturdy cast aluminum alloy clamp can be swiveled and locked at any desired angle.
The clamp can be attached to rods up to 13 mm ( 1/2") in diameter.
Specifications
  • Description:
    Adjustable Right Angle Clamp Holder