Orders LinkContactUsLinkComponent
Conductivity Indicator
Conductivity Indicator
Supplier:   Science First
Conductivity Indicator
Supplier:  Science First
Certificates
Conductivity Indicator
Conductivity Indicator
2 Options Available Below
Product Details & Documents
Light Bulb Glows When Water Conducts Electricity.
  • Attaches to a standard ring stand
  • Comes with illustrated instructions
Presenting no shock hazard because the cord at the side can’t come in contact with the water, this indicator features a plastic molded body and recessed electrical connections and electrodes. It operates on 110 VAC. A light bulb is required.
Product Family Options
Product Information
  • Availability
    Price
  • 470016-796
    Conductivity Indicator
    Conductivity Indicator
    Log in for availability
    Log in for availability
    Each
    Each - $73.05
    $73.05
    Each
       $73.05
    Supplier Number: 611-2260
  • 470061-754
    Bulb (15W) for Water Conductivity
    Bulb (15W) for Water Conductivity
    Log in for availability
    Log in for availability
    Each
    Each - $15.60
    $15.60
    Each
       $15.60
    Supplier Number: 674-0035