1" Square Color Tiles
Catalog # 470145-598
Supplier: Skywill Far East Industries
Specifications
- Description:1" Square Color Tiles, 0.5cm Thick0
- Cat. No.:470145-598
- Supplier no.:M1330
Specifications
About this item
An excellent item for patterning, teaching area, perimeter, counting and sorting, these 1"x1" tiles come in four bright colors.
- Excellent for patterning
- Bright colors
Tiles are 0.5cm thick. Pack of 400.
Caution: WARNING: CHOKING HAZARD - Small Parts. Not for children under 3 years.