Orders LinkContactUsLinkComponent
Geraniol for synthesis
Geraniol for synthesis
Geraniol for synthesis
Geraniol for synthesis
Geraniol, a monoterpenoid, occurs in small quantities in lemon, geranium, and other essential oils. It is used in flavors like raspberry and plum. It is also used in plant-based mosquito repellent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
 :  (E)-3,7-Dimethylocta-2,6-dien-1-ol
  •  :106-24-1
  •  :C₁₀H₁₈O
  •  :MFCD00002917
  •  :154.25 g/mol
  •  :0.877
  •  :Ambient
  •  :203-377-1
Documents
  • Assay:
    96% Min.
Product Family Options
  • Pack type
    Size
  • Glass, amber
    25 g
    Each (25g)
    Each (25g) - $93.49
    $93.49
    Each (25g)
       $93.49
     : G1008-25GM
  • Glass, amber
    100 g
    Each (100g)
    Each (100g) - $164.73
    $164.73
    Each (100g)
       $164.73
     : G1008-100GM
  • Poly pail
    5 kg
    Each (5kg)
    Each (5kg) - $5,234.87
    $5,234.87
    Each (5kg)
       $5,234.87
     : G1008-5KGBL