Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Polydimethylsiloxane, Technical Grade for synthesis

Polydimethylsiloxane, Technical Grade for synthesis
Certificates
Polydimethylsiloxane, Technical Grade for synthesis

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Dimethylpolysiloxane, Technical, also known as polydimethylsiloxane, is a silicon-based surfactant used in defoamers for over-the-counter drugs and personal care products.

Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use.

Synonyms:  Dimethylpolysiloxane, PDMS, Silicone oil, Dimethicone
  • CAS Number:63148-62-9
  • Formula:(CH₃)₃SiO(Si(CH₃)₂O)n
  • MDL Number:MFCD00084411
  • Storage Temperature:Ambient

Documents

Certificates
Search by Lot Number
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass, amber
    100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $110.29
    $110.29
    Each (100g)
       $110.29

    Supplier Number: D1228-100GM
  • Glass, amber
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $318.53
    $318.53
    Each (500g)
       $318.53

    Supplier Number: D1228-500GM
  • Glass bottle, poly-coated
    2.5 kg
    Log in for availability
    Log in for availability
    Each (2,500g)
    Each (2,500g) - $1,032.06
    $1,032.06
    Each (2,500g)
       $1,032.06

    Supplier Number: D1228-2.5KG

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.