Adapter, Thermocouple Well, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
This adapter features a ground inner standard taper joint for adapting thermocouples into jointed heads.
- Ground inner [ST] joint
- Adapts thermocouples into jointed heads
- Fabricated from borosilicate glass
Adapter well is fabricated from very thin wall borosilicate glass to allow for better temperature transfer. The well is fabricated in two lengths, for 25mm or 76mm immersion. Length is measured below the joint. The 10/30 joint version is for Micro/Mini-Lab scale use.