Adapter, Stopcock, Ace Glass Incorporated
89074-290
Product available on GSA Advantage®
Small Business Enterprise
- Description:Vacuum Adapter, with Stopcock
- Cat. No.:89074-290
- Supplier no.:9175-04
This vacuum adapter features a 2mm bore glass stopcock on the side arm.
- Side arm with glass stopcock
- Top ground [ST] outer joint
- Bottom ground [ST] inner joint
- Fabricated from borosilicate glass
Adapter has a ground 14/20 standard taper outer joint at the top and a ground standard taper 14/20 inner joint at the bottom.