ContactUsLinkComponent
Beakers, Jacketed, Ace Glass Incorporated
Beakers, Jacketed, Ace Glass Incorporated
Catalog # 89082-884
Supplier:  Ace Glass
Supplier #: 5340-135
Product available on GSA Advantage®
Small Business Enterprise
undefined
Beakers, Jacketed, Ace Glass Incorporated
Catalog # 89082-884
Supplier:  Ace Glass
Supplier Number:  5340-118
Supplier #: 5340-135
Product available on GSA Advantage®
Small Business Enterprise
Log in for availability
Select an Option
160 mm (I.D.)
Select an Option
5 L (169 oz.)
Select an Option
Jacketed Beaker with Ace-Safe Connectors
Select an Option
250 mm
Select an Option
89082-884
Select an Option
89052-958 (For Tube Size<sup>1</sup>/<sub>2</sub>")
Select an Option
5340-135
About this item
This jacketed beaker has one upper and one lower Ace-Thred connection on opposite sides.
Does not have a pouring spout. Fabricated from borosilicate glass.