ContactUsLinkComponent
Erem® Cutting Swiss Tweezers, Cooper Tools
Erem® Cutting Swiss Tweezers, Cooper Tools
Catalog # 37691-628
Supplier:  Cooper Tools
undefined
Erem® Cutting Swiss Tweezers, Cooper Tools
Catalog # 37691-628
Supplier:  Cooper Tools
Supplier Number:  15AGW
Specifications
  • Material:
    Carbon steel
  • Environmentally Preferable:
  • Cat. No.:
    37691-628
  • Cutting capacity:
    ½" soft wire
  • Tip style:
    Curved
  • Overall length:
    11.4 cm (4½")
  • Supplier no.:
    15AGW
Specifications
About this item
Oblique heat cutting tweezers for cutting soft wire. Induction hardened cutting blades improves cutting capability and lifetime.
  • Made of tempered carbon steel
  • Anti-glare finish
This is a llong, anti-acid, medium curved tip tweezer and is perfect for use in micro-electronic applications.