NMC Single Entry Plug Lockout
Catalog # 94016-488
Supplier: ACCUFORM MANUFACTURING, INC
Supplier #: LP110
Product Details & Documents
Accommodates plugs up to 4.45cm (1¾") dia., 8.26cm (3¼") long with a maximum cord dia
Of 1.27cm (½"). Accepts 110 volt and certain models of 220 volt plugs.