Order Entry
Puerto Rico
ContactUsLinkComponent
 
Adapter, [SJ], Angled Hose Connection, Ace Glass Incorporated
  89051-396
 :  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Adapter, [SJ], Angled Hose Connection, Ace Glass Incorporated
  89051-396
 :  Ace Glass
 :  5200-35
Product available on GSA Advantage®
Small Business Enterprise

 

  • [ST] joint:
    35/25
  • Description:
    Glass Stopcock Straight Hose Connection Adapter
  • Cat. No.:
    89051-396
  • Bore size:
    3 mm
  • Hose connections:
    5/16" or 3/8" (C)
  • Supplier no.:
    5200-35

 

 

This adapter features a 90 degree angled hose connection with a spherical ball joint at the bottom and glass stopcock.

  • 90° angled hose connection
  • Bottom spherical ball joint
  • Glass stopcock
  • Fabricated from borosilicate glass

Adapter has a glass stopcock with a 3mm plug bore. Bore size represents the plug bore size.