ContactUsLinkComponent
Adapter, Hose Connection, Ace Glass Incorporated
Adapter, Hose Connection, Ace Glass Incorporated
  89051-418
 :  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Adapter, Hose Connection, Ace Glass Incorporated
  89051-418
 :  Ace Glass
 :  5215-10
Product available on GSA Advantage®
Small Business Enterprise
  • [ST] joint:
    24/40
  • Description:
    Hose Connection Adapter
  • Cat. No.:
    89051-418
  • Supplier no.:
    5215-10
This adapter features a reinforced standard taper outer joint and a hose connection.
  • Reinforced [ST] outer joint
  • With hose connection
  • Fabricated from borosilicate glass
Use adapter with a 3/8-inch I.D. tubing, size D hose connection.