Adapter, Adjustable Flow Stopcock, Ace Glass Incorporated
Catalog # 89051-458
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Adjustable Flow Stopcock Adapter
- Cat. No.:89051-458
- Supplier no.:5250-10
Specifications
About this item
This adapter features an adjustable flow glass stopcock.
- Glass stopcock
- Top reinforced [ST] outer joint
- Bottom [ST] inner joint
- Fabricated from borosilicate glass
Featuring a reinforced 24/40 standard taper outer joint at top and 24/40 standard taper inner joint at bottom.