Order Entry
Puerto Rico
ContactUsLinkComponent
Adapter, Jacketed, Ace Glass Incorporated
Adapter, Jacketed, Ace Glass Incorporated
  89051-296
 :  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Adapter, Jacketed, Ace Glass Incorporated
  89051-296
 :  Ace Glass
 :  5155-10
Product available on GSA Advantage®
Small Business Enterprise

 

  • Description:
    105 Degree Angle Jacketed Adapter
  • Cat. No.:
    89051-296
  • Supplier no.:
    5155-10

 

 

This adapter is jacketed with water-cooled 24/40 standard taper joints.

  • Extension arm below joint
  • 105° angle
  • Jacketed
  • Fabricated from borosilicate glass

The extension arm below the joint is 50mm. Use with a 3/8-inch I.D. tubing, size D hose connection.