Adapter, 'U', [ST] 24/40, Ace Glass
Catalog # 89051-260
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:"U" Adapter
- Environmentally Preferable:
- Cat. No.:89051-260
- Supplier no.:5060-10
Specifications
About this item
This is a connecting adapter with reinforced standard taper 24/40 outer joints at both ends.
- U-shaped
- Reinforced [ST] outer joints at both ends
- Fabricated from borosilicate glass