Brushes, Bench, Wooden Handle, Excelta Corp®
Supplier: Avantor
Certificates
About this item
Bench brushes are ideal for general cleaning large areas.
- For large areas
- Static-safe bristles
These bench brushes are anti-static (10⁸-10¹¹ ohms/sq) and are made of high grade bristles with a wooden handle. They are ideal for large scale cleaning.