Instatherm® Economy Bath/Controller Kit, Ace Glass Incorporated
Catalog # 89075-340
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Instatherm® Economy Bath/Controller Kit
- Cat. No.:89075-340
- Supplier no.:9601-335
Specifications
About this item
Instatherm® is a safer, more precise heating method for the laboratory
This economy bath/controller kit consists of one 89075-328 bath with integral six-foot controller cord, one 89076-514 economy temperature controller, one 89075-336 clip, and one 89076-450 type "J" thermocouple temperature sensor and controller cord.
It is an Ace Glass proprietary process that fuses a noble metal alloy coating directly onto the glass surface. Once the insulating and protective covering is applied, the Instatherm® electrical circuit provides the most rapid and precise heating method available. Thermal lag is minimal with no hot spots. When placed on a magnetic stirrer, it becomes the best heating and mixing combination on the market.
Ordering information: Because Instatherm® is a process more than product, it can be adhered to any glass vessel. For more information on custom applications, contact your Avantor representative.