Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Tryptone Base Materials

Tryptone Base Materials

Supplier:   G-Biosciences
Tryptone Base Materials
Supplier:  G-Biosciences
Certificates
Tryptone Base Materials

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Tryptone is an assortment of peptides generated by the tryptic digestion of casein. It is commonly used in microbiology to produce Lysogeny broth for the growth of E. coli and other microorganisms, as it provides a source of amino acids.

  • Solubility: Water

  • Appearance: Cream to beige free flowing powder
  • CAS: 91079-40-2

Documents

Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Tryptone
    Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    Each
    Each - $62.65
    $62.65
    Each
       $62.65

    Supplier Number: RC-109
  • Tryptone
    Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    Each
    Each - $214.82
    $214.82
    Each
       $214.82

    Supplier Number: RC-110
  • 76286-950
    Tryptone
    Tryptone
    Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    Each
    Each - $98.95
    $98.95
    Each
       $98.95

    Supplier Number: RC-111
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.