Specifications
- Description:Immobilized D-Galactose
- Size:5 mL
- Cat. No.:71003-140
- Supplier no.:786-391
Specifications
About this item
G-Biosciences' Immobilized D-Galactose is designed for the rapid purification and enrichment of glycoproteins, polysaccharides, glycolipids, lectins, galactosidases and other galactose-binding molecules.
G-Biosciences' Immobilized D-Galactose is also ideal for the purification of agglutinins, lectins, toxins, galactose-binding, N-acetylgalactosamine-binding or carbohydrate binding molecules.
G-Biosciences' Immobilized D-Galactose consists of agarose covalently coupled to D-galactose. The binding capacity of Immobilized D-Galactose is >20 mg Jacalin per mL resin.