Order Entry
United States
ContactUsLinkComponent
Pearl™ Monoclonal IgG Purification, G-Biosciences
Pearl™ Monoclonal IgG Purification, G-Biosciences
Catalog # 89168-060
Supplier:  G-Biosciences
CAS Number:  
Pearl™ Monoclonal IgG Purification, G-Biosciences
Catalog # 89168-060
Supplier:  G-Biosciences
Supplier Number:  786-802
CAS Number:  

Specifications

  • Description:
    Pearl™ Monoclonal IgG Purification Kit
  • Size:
    1 kit
  • Environmentally Preferable:
  • Cat. No.:
    89168-060
  • Supplier no.:
    786-802

Specifications

About this item

G-Biosciences' Pearl™ Monoclonal IgG Purification kit allows for the rapid purification of antibodies from cell culture supernatant and ascites fluid

G-Biosciences' Pearl™ Monoclonal IgG Purification kit can be used to purify antibodies direct from cell culture supernatant with less than 10% FBS or can be used with ascites fluid after treatment with the supplied Ascites PreTreat.

G-Biosciences' Pearl™ Monoclonal IgG Purification kit can purify IgG from ~1L cell culture supernatant or 200 mL ascites fluid.

The Pearl™ IgG Purification Resin binds the high abundant, non-IgG proteins (i.e. albumin) and allows the IgG molecules to pass through in a physiological buffer. The IgG molecules can be stored or used in downstream applications without further clean-up, such as ammonium sulfate precipitation.

Ordering information: Accessories included. Additional supplies available.