ContactUsLinkComponent
Biotin Conjugation Estimation, G-Biosciences
Biotin Conjugation Estimation, G-Biosciences
Catalog # 82023-262
Supplier:  G-Biosciences
CAS Number:  
Biotin Conjugation Estimation, G-Biosciences
Catalog # 82023-262
Supplier:  G-Biosciences
Supplier Number:  BKC-01
CAS Number:  
Specifications
  • Description:
    HOOK™ BiotinQuant Kit
  • Size:
    20 assays
  • Cat. No.:
    82023-262
  • Supplier no.:
    BKC-01
Specifications
About this item
G-Biosciences' HOOK™ BiotinQuant is designed for the estimation of biotin conjugation.
G-Biosciences' HOOK™ BiotinQuant™ uses HABA [4’-hydroxyazobenzene-2-carboxylic acid] dye to rapidly estimate the mole-to-mole ratio of biotin to antibody or protein. The method of biotin incorporation estimation is based on the binding of avidin with HABA dye, which produces a color that can be read at 500 nm. The HABA-avidin complex can be displaced with free biotin or biotin conjugated with other molecules (proteins). Measuring the change in optical density of HABA-avidin complex with biotinylated proteins allows for accurate estimation of the molar ration of biotin conjugated to the protein/ antibody.
G-Biosciences' HOOK™ BiotinQuant™ is supplied with premixed, OneQuant™ HABA/Avidin aliquots to eliminate difficulties in weighing small amounts of reagents and ensuring reproducible estimations.
Ordering information: Accessories included. Additional supplies available.