Order Entry
Puerto Rico
ContactUsLinkComponent
Copper Chelating Resin for Immobilized Metal Affinity Chromatography Purification, G-Biosciences
Copper Chelating Resin for Immobilized Metal Affinity Chromatography Purification, G-Biosciences
Catalog # 89131-008
Supplier:  G-Biosciences
CAS Number:  
Copper Chelating Resin for Immobilized Metal Affinity Chromatography Purification, G-Biosciences
Catalog # 89131-008
Supplier:  G-Biosciences
Supplier Number:  786-285
CAS Number:  

Specifications

  • Description:
    Copper Chelating Resin
  • Size:
    10 mL Resin
  • Cat. No.:
    89131-008
  • Supplier no.:
    786-285

Specifications

About this item

G-Biosciences' Copper Chelating Resin is an immobilized metal affinity chromatography (IMAC) resin that was specifically designed to utilize Cu²⁺ for the purification of copper binding proteins, including 6x histidine tagged proteins.

G-Biosciences' Copper Chelating Resin binds to six histidine residues (6x His), a common tag used in protein purification. The resin consists of iminodiacetate coupled to 6% cross-linked agarose beads. The iminodiacetate binds divalent copper ion with a capacity of 20-40 μmoles Cuu²⁺/mL resin. The protein binding capacity is >50 mg protein per ml resin. We have demonstrated binding of >100 mg of a 50 kDa 6X His tagged proteins to a mL of resin.

G-Biosciences' Immobilized Nickel, Cobalt and Zinc Chelating Resins are also available. Cobalt has the highest selectivity followed by Zinc, Nickel then Copper, but has the lowest loading capacity. Copper has the highest loading capacity, followed by Nickel then Zinc. The capacity of copper is >50 mg per mL. Specific binding/wash and elution buffers are available.

Ordering information: Accessories available.