ContactUsLinkComponent
SDC™ (Steroid/Drug/Compound) Immobilization Kit, G-Biosciences
SDC™ (Steroid/Drug/Compound) Immobilization Kit, G-Biosciences
  71003-104
 :  
undefined
SDC™ (Steroid/Drug/Compound) Immobilization Kit, G-Biosciences
  71003-104
 :  786-271
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    SDC™ (Steroid/Drug/Compound) Immobilization
  • Size:
    For 5 columns
  • Cat. No.:
    71003-104
  • Supplier no.:
    786-271
G-Biosciences' SDC™ (Steroid/ Drug/ Compound) Immobilization kit is designed for the immobilization of steroids, drugs and chemical compounds that lack primary amines, sulfhydryls, carbonyls and other common coupling groups to a solid-phase agarose support for the use in affinity purification
G-Biosciences' SDC™ kit uses the Mannich reaction for coupling, which is described as the condensation of formaldehyde with ammonia, in the form of its salt, and another compound containing an active hydrogen. The SDC™ Immobilization kit replaces the ammonia with the primary amine on the DADPA and the active hydrogen is supplied by the steroid, drug or chemical compound to be coupled.
G-Biosciences' SDC™ kit is ideal for the generation of five 2 mL affinity columns.
The kit uses our Immobilized DADPA (diaminodipropylamine) resin that can bind steroids, drugs and chemical compounds through their active hydrogens.
 : Accessories included. Additional supplies available.