About this item
G-Biosciences' GeneCAPSULE™ is an electro-elution tool for rapid, nearly 90% recovery of PCR products, DNA fragments and proteins from agarose and polyacrylamide gels
Electroelution through GeneCAPSULE™ is a gentle method that eliminates the risks of damage to DNA or protein samples commonly encountered in use of glass milk, spin columns, binding columns and other popular techniques. The GeneCAPSULE™ method is simple to perform and eliminates additional steps involving washing, spinning, heating or precipitation etc. The recovered DNA is of highest quality for use in molecular manipulations such as ligation, restriction enzyme digestion, sequencing, amplification, random priming, and other enzymatic reactions.
G-Biosciences' G-CAPSULE™ can be used for extraction of >20bp DNA and RNA or for >4kDa proteins. The G-CAPSULE™ Weight is a small weight device that prevents G-CAPSULE™ from floating during electroelution.
G-CAPSULE™ consists of mainly two parts, Gel-PICK™ - for picking DNA or protein band from the gel, and the Gene-TRAP™, which traps the migrating DNA or protein during electroelution.
Kit Electroelution includes agarose, DNA stain (500X), DNA ladder (1kb), DNA loading buffer (6X), TAE buffer (50X), centrifuge tubes (2ml), GeneCAPSULE electroelution devices.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Method / Format:Electroelution
- Prep size:Scalable
- Refine target molecule:RNA gel bandsDNA gel bandsProtein gel bands
- Sample type:Gel matrix
- Target molecule:DNARNAProtein
- Description:GeneCAPSULE™ Electroelution device