ContactUsLinkComponent
Fast Yeast Transformation™ for Rapid Generation of Competent Yeast Cells, G-Biosciences
Fast Yeast Transformation™ for Rapid Generation of Competent Yeast Cells, G-Biosciences
Catalog # 82023-072
Supplier:  G-Biosciences
Fast Yeast Transformation™ for Rapid Generation of Competent Yeast Cells, G-Biosciences
Catalog # 82023-072
Supplier:  G-Biosciences
Supplier Number:  GZ-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Fast Yeast Transformation™
  • Size:
    120 preps
  • Cat. No.:
    82023-072
  • Supplier no.:
    GZ-1
Specifications
About this item
G-Biosciences' Fast Yeast Transformation™ kit is a rapid, single-step, yeast transformation kit that generates competent yeast cells and plasmids suitable for multiple downstream applications.
G-Biosciences' Fast Yeast Transformation™ kit prepares competent yeast cells in less than 10 minutes. The competent yeast cells can be used immediately or frozen (for up to six months) for later use. This method is suitable for both circular and linear plasmid transformations. The protocol involves simply suspending yeast cells in the supplied Competent Buffer, and the cells are then ready to receive transforming DNA. Introduce DNA to the competent cells and incubate for transformation.
G-Biosciences' Fast Yeast Transformation™ kit is compatible with C. albicans, S. pombe, P. pastoris, or S. cerevisiae. High transformation efficiency: 10⁵-10⁶ transformants/μg circular DNA.
Ordering information: Accessories available.