Anti-SARS virus Spike Rabbit Polyclonal Antibody
Catalog # 80088-820
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:SARS virus Spike
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:SARS virus
- Size:100 µg
- Epitope:N-Terminal
- Cat. No.:80088-820
- Supplier no.:AP6009B
Specifications
About this item
Primary Rabbit Anti-SARS virus Spike (N-term D204) Reacts with SARS Virus
Type: Primary
Antigen: SARS virus Spike (N-term D204)
Clonality: Polyclonal
Clone:
Conjugation: (none)
Epitope:
Host: Rabbit
Isotype:
Reactivity: SARS Virus