Anti-SARS virus Spike Rabbit Polyclonal Antibody
80088-828
: Abgent
- Antibody Type:Primary
- Antigen Name:SARS virus Spike
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:SARS virus
- Size:100 µg
- Epitope:C-Terminal (V1158, 1150–1200 aa)
- Cat. No.:80088-828
- Supplier no.:AP6009G
Primary Rabbit Anti-SARS virus Spike (C-term V1158) Reacts with SARS Virus
Type: Primary
Antigen: SARS virus Spike (C-term V1158)
Clonality: Polyclonal
Clone:
Conjugation: (none)
Epitope:
Host: Rabbit
Isotype:
Reactivity: SARS Virus