Anti-SARS virus PUPM Rabbit Polyclonal Antibody
80088-816
: Abgent
- Antibody Type:Primary
- Antigen Name:SARS virus PUPM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:SARS virus
- Size:100 µg
- Epitope:N-Terminal
- Cat. No.:80088-816
- Supplier no.:AP6008A
Primary Rabbit Anti-SARS virus PUPM (N-term) Reacts with SARS Virus
Type: Primary
Antigen: SARS virus PUPM (N-term)
Clonality: Polyclonal
Clone:
Conjugation: (none)
Epitope:
Host: Rabbit
Isotype:
Reactivity: SARS Virus