Anti-PI3KCB Rabbit Polyclonal Antibody
Catalog # 80084-686
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit beta isoform
- Antigen Symbol:PI3KCB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Size:100 µg
- Epitope:N-Terminal (151–165 aa)
- Antigen Synonyms:beta isoform|PIK3CB|Phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit|PI3-kinase p110 subunit beta
- Cat. No.:80084-686
- Supplier no.:AP8017A
Specifications
About this item
Primary Rabbit Anti-PI3KCB (N-term) Reacts with Human
Type: Primary
Antigen: PI3KCB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal (151–165 aa)
Host: Rabbit
Isotype:
Reactivity: Human