Anti-MAGEA5 Rabbit Polyclonal Antibody
Catalog # 80088-978
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Melanoma Antigen Family A5
- Antigen Symbol:MAGEA5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Size:100 µg
- Epitope:N-Terminal (1–100 aa)
- Antigen Synonyms:MAGE5|MAGEA4|CT1.5
- Cat. No.:80088-978
- Supplier no.:AP6167A
Specifications
About this item
Primary Rabbit Anti-MAGEA5 (N-term) Reacts with Human
Type: Primary
Antigen: MAGEA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal (1–100 aa)
Host: Rabbit
Isotype:
Reactivity: Human