To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin L
- Antigen Symbol:IgL
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Size:1 mg
- Environmentally Preferable:
- Epitope:Light Chain
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Antigen Synonyms:lambda light chain|Ig lambda|Ig lambda light chain|Immunoglobulin lambda light chain|Igl
- Storage Buffer:0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
- Storage Temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Ambient
- Immunogen:Mouse lambda light chain
- Tested Applications:Fluoresence ELISA, Fluorescence Western Blot
- Purification:purified by immunoaffinity chromatography
- Cat. No.:RL610-4211
- Supplier no.:610-4211
Secondary Rabbit Anti-λ (lambda chain) Reacts with Mouse
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgL
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Light Chain
Host: Rabbit
Isotype: IgG
Reactivity: Mouse