To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Tentoxylysin
- Antigen Symbol:TETX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Clostridium tetani
- Western Blot:Yes
- Size:100 µL
- Epitope:Full length
- Cross Adsorption:No
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Gene ID:1061100
- Antigen Synonyms:TeNT-LC|tetX|tetanus toxic fragment C|Tentoxylysin
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide
- Storage Temperature:–20 °C
- Concentration:63 mg/ml by Refractometry
- Shipping Temperature:Dry Ice
- Immunogen:This antibody was prepared from whole rabbit serum produced by repeated immunizations with a polyhistidine-tagged TTFC fusion Protein expressed in E. coli corresponding to full length TTFC.
- Purification:delipidation and defibrination
- Cat. No.:RL100-401-894
- Supplier no.:100-401-894
This antibody has been tested for use in ELISA and western blot. For western blots expect a band of approximately 52 kDa in size corresponding to full-length TTFC. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: ELISA: 1:50000 - 1:250000; Western Blot: 1:1000 - 1:40000;
Type: Primary
Antigen: tetX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Clostridium tetani