Anti-SARS-CoV Nucleocapsid Rabbit Polyclonal Antibody
RL200-401-A50
- Antibody Type:Primary
- Antigen Name:SARS-CoV Nucleocapsid
- Antigen Symbol:SARS-CoV Nucleocapsid
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:SARS virus
- Western Blot:Yes
- Size:500 µg
- Epitope:Full length
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Gene ID:1489678
- Antigen Synonyms:SARS-CoV Nucleocapsid Protein
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage Temperature:–20 °C
- Concentration:5.0 mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Ambient
- Immunogen:This Protein A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a purified recombinant Protein corresponding to full length SARS Coronavirus Nucleocapsid Protein. Lifesensors Inc. (www.lifesensors.com) prepare
- Purification:Protein A purified
- Cat. No.:RL200-401-A50
- Supplier no.:200-401-A50
This Protein A purified antibody has been tested for use in ELISA and by western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 46 kDa in size corresponding to SARS Nucleocapsid (N) Protein.
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:2000 - 1:10000
Restore with deionized water (or equivalent); 500 µl
Type: Primary
Antigen: SARS-CoV Nucleocapsid
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: SARS Virus