Anti-NSP13 Rabbit Polyclonal Antibody [clone: 5D11.G5]
RL100-401-A54
- Antibody Type:Primary
- Antigen Name:SARS-CoV Nonstructural Protein 13
- Antigen Symbol:NSP13
- Clonality:Polyclonal
- Clone:5D11.G5
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:SARS virus
- Western Blot:Yes
- Size:100 µL
- Epitope:C-Terminal
- Cross Adsorption:No
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Gene ID:1489680
- Antigen Synonyms:SARS-CoV Nonstructural Protein 13
- Amino Acid Number:584 to 601
- Storage Buffer:0.01% (w/v) Sodium Azide
- Storage Temperature:–20 °C
- Concentration:85 mg/ml by Refractometry
- Shipping Temperature:Dry Ice
- Immunogen:This antibody was prepared from whole rabbit serum produced by repeated immunizations with a BSA-coupled synthetic peptide corresponding to the C-terminus (amino acid residues 584-601) of the SARS Coronavirus nonstructural Protein 13.
- Tested Applications:Immunoelectron Microscopy
- Cat. No.:RL100-401-A54
- Supplier no.:100-401-A54
This antibody has been tested for use in western blotting, immunofluorescence microscopy and immunoelectron microscopy. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 67 kDa in size.
Recommended Dilutions: Immunofluorescence: 1:800; Western Blot: 1:1000; Immunoprecipitation: 1:60; Immunoelectron Microscopy 1:300
Type: Primary
Antigen: NSP13
Clonality: Polyclonal
Clone: 5D11.G5
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: SARS Virus