Anti-RBC Rabbit Polyclonal Antibody [clone: BM3]
Catalog # RL112-4139
Supplier: Rockland Immunochemical
Specifications
- Antibody Type:Primary
- Antigen Name:Red Blood Cell antigen
- Antigen Symbol:RBC
- Clonality:Monoclonal
- Clone:BM3
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Rat
- Size:2 mL
- Cross Adsorption:No
- Format:Antiserum
- Form:Lyophilized
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.20.01% (w/v) Sodium Azide
- Storage Temperature:–20 °C
- Concentration:90 mg/ml by Refractometry
- Shipping Temperature:Ambient
- Immunogen:Rat washed pooled Red Blood Cells (RBC)
- Tested Applications:Agglutination
- Cat. No.:RL112-4139
- Supplier no.:112-4139
Specifications
About this item
Primary Rabbit Anti-RAT Red Blood Cell (RBC) Reacts with Rat
Recommended Dilutions: Agglutination Titer 1:32 - 1:64;
Restore with deionized water (or equivalent); 2.0 ml
Type: Primary
Antigen: RBC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat