To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:RING-box Protein 1
- Antigen Symbol:RBX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross Adsorption:No
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Gene ID:9978
- Antigen Synonyms:RNF75|BA554C12.1|ROC1|E3 Ubiquitin Protein Ligase|Ring-box 1
- Amino Acid Number:94 to 108
- Storage Buffer:0.01% (w/v) Sodium Azide
- Storage Temperature:–20 °C
- Concentration:85 mg/ml by Refractometry
- Shipping Temperature:Dry Ice
- Immunogen:This antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 94-108 of Human ROC1 (C-terminal) coupled to KLH.
- Purification:delipidation and defibrination
- Cat. No.:RL100-401-A13
- Supplier no.:100-401-A13
Specifications
About this item
This antibody reacts with human ROC1 by western blot and immunoprecipitation. The antibody immunoprecipitates in vitro translated Protein and Protein from overexpressing cell lysates (using HeLa and NIH-3T3, and others).
Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;
Type: Primary
Antigen: RBX1
Clonality: Monoclonal
Clone: 14A11
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human