Order Entry
United States
ContactUsLinkComponent
Anti-IgM µ Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anti-IgM µ Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # RL610-4507
undefined
Anti-IgM µ Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # RL610-4507
Supplier Number:  610-4507
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin M mu-chain
  • Antigen Symbol:
    IgM µ
  • Clonality:
    Polyclonal
  • Conjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Mouse
  • Western Blot:
    Yes
  • Size:
    1 mg
  • Environmentally Preferable:
  • Epitope:
    µ-chain
  • Cross Adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Storage Buffer:
    0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0, 0.1% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
  • Storage Temperature:
    +4 °C
  • Concentration:
    1.0 mg/ml by UV absorbance at 280 nm
  • Shipping Temperature:
    Wet Ice
  • Immunogen:
    Mouse IgM mu heavy chain
  • Tested Applications:
    Dot Blot
  • Purification:
    purified by immunoaffinity chromatography
  • Cat. No.:
    RL610-4507
  • Supplier no.:
    610-4507

Specifications

About this item

Anti-MOUSE IgM (alpha chain) (RABBIT) Antibody is suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:20000;
Do not freeze!

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: µ-chain
Host: Rabbit
Isotype: IgM
Reactivity: Mouse