AKT/PKBa Substrate Peptide
Catalog # RL000-000-260
Supplier: Rockland Immunochemical
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Conjugation:Unconjugated
- Size:500 µg
- Protein/Peptide Name:AKT/PKBa Substrate Peptide
- Molecular Weight:817 kDa
- Sequence:RPRAATF
- Tested Applications:ELISA, Flow Cytometry, ImmunoChemistry, ImmunoPrecipitation, Western Blot
- Cat. No.:RL000-000-260
- Supplier no.:000-000-260
Specifications
About this item
Primary Anti-AKT/PKBa Reacts with Pan