Anti-ABL2 Rabbit Polyclonal Antibody
Catalog # 80084-360
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:ABL proto-oncogene 2, Non-receptor Tyrosine Kinase
- Antigen Symbol:ABL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Size:100 µg
- Epitope:N-Terminal (1–100 aa)
- Antigen Synonyms:ARG|ABL2|FLJ22224|FLJ41441|FLJ31718|ABLL
- Cat. No.:80084-360
- Supplier no.:AP7695A
Specifications
About this item
Primary Rabbit Anti-ABL2 (N-term) Reacts with Human
Type: Primary
Antigen: ABL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal (1–100 aa)
Host: Rabbit
Isotype:
Reactivity: Human