Nalgene® Oak Ridge Conical Centrifuge Tube Adapter; white PC, Thermo Scientific
21010-613
- Cat. No.:21010-613
- Packaging:6 Per Pkg.
- Supplier no.:DS3147-0050
Nalgene oak ridge conical tube adapter must be used with Nalgene 35 L conical oak ridge tubes.